Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Ptp4a3
- Entrez Gene ID
-
19245
- Gene Name
- protein tyrosine phosphatase 4a3
- Gene Aliases
-
AV088979, Prl-3, pPtp4a3
- Chromosome Location
-
Chr.15: 73723145 - 73758766 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK004562.1 |
- |
1 - 2 |
55 |
68 |
View Full Product Details