Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Uevld
Entrez Gene ID
54122
Gene Name
UEV and lactate/malate dehyrogenase domains
Gene Aliases
8430408E05Rik, Attp
Chromosome Location
Chr.7: 46923216 - 46958530 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001040695.1 NP_001035785.1 2 - 3 212 80
GenBank mRNA AK154536.1 - 2 - 3 163 80
AK155685.1 - 2 - 3 159 80
BC054796.1 - 2 - 3 218 80
CB524764.1 - 2 - 3 218 80
CN538811.1 - 1 - 2 82 80
U97571.1 - 2 - 3 166 80

View Full Product Details