Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Apba1
Entrez Gene ID
319924
Gene Name
amyloid beta (A4) precursor protein binding, family A, member 1
Gene Aliases
6430513E09Rik, Lin-10, Mint, Mint1, Urop11, X11, X11alpha
Chromosome Location
Chr.19: 23758876 - 23949598 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_177034.3 NP_796008.2 4 - 5 1642 138
GenBank mRNA AK032261.1 - 3 - 4 406 138
AK147583.1 - 4 - 5 1632 138
AK147623.1 - 4 - 5 1591 138
BC141181.1 - 4 - 5 1460 138

View Full Product Details