Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Med25
Entrez Gene ID
75613
Gene Name
mediator complex subunit 25
Gene Aliases
2610034E13Rik, 2610529E18Rik, ESTM2
Chromosome Location
Chr.7: 44876765 - 44892664 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_029365.2 NP_083641.1 2 - 3 183 64
GenBank mRNA AK089940.1 - 2 - 3 184 64
AK090026.1 - 2 - 3 209 64
AK148368.1 - 2 - 3 236 64
AK149867.1 - 2 - 3 224 64
AK158238.1 - 2 - 3 219 64
AK165760.1 - 2 - 3 228 64
BC021333.1 - 2 - 3 218 64
BC031138.1 - 2 - 3 226 64

View Full Product Details