Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Med25
Entrez Gene ID
75613
Gene Name
mediator complex subunit 25
Gene Aliases
2610034E13Rik, 2610529E18Rik, ESTM2
Chromosome Location
Chr.7: 44876765 - 44892664 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_029365.2 NP_083641.1 7 - 8 816 92
GenBank mRNA AK089940.1 - 7 - 8 820 93
AK090026.1 - 7 - 8 842 92
AK149867.1 - 8 - 9 1088 92
AK158238.1 - 7 - 8 877 92
AK165760.1 - 7 - 8 862 92
BC021333.1 - 7 - 8 851 92
BC031138.1 - 7 - 8 859 92
BC071206.1 - 6 - 7 893 92

View Full Product Details