The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001109909.1 | NP_001103379.1 | 7 - 8 | 1211 | 76 |
NM_001109910.1 | NP_001103380.1 | 7 - 8 | 1211 | 76 | |
NM_031405.2 | NP_113582.1 | 7 - 8 | 1211 | 76 | |
GenBank mRNA | AK144300.1 | - | 7 - 8 | 1202 | 76 |
BC066831.1 | - | 7 - 8 | 1170 | 76 |