Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Sult2a6
Entrez Gene ID
629219
Gene Name
sulfotransferase family 2A, dehydroepiandrosterone (DHEA)-preferring, member 6
Gene Aliases
EG629219, Gm6957
Chromosome Location
Chr.7: 14222403 - 14254870 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001081325.2 NP_001074794.1 3 - 4 513 151
GenBank mRNA BC147210.1 - 3 - 4 513 151

View Full Product Details