Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
N4bp2
Entrez Gene ID
333789
Gene Name
NEDD4 binding protein 2
Gene Aliases
B3bp, E430014I16, Gm1791, Gm868
Chromosome Location
Chr.5: 65761726 - 65830107 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001024917.1 NP_001020088.1 18 - 19 5417 109
GenBank mRNA AK136182.1 - 8 - 9 2564 109
CK637074.1 - 3 - 4 384 109
CX769144.1 - 2 - 3 362 109

View Full Product Details