Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Agpat6
Entrez Gene ID
102247
Gene Name
1-acylglycerol-3-phosphate O-acyltransferase 6 (lysophosphatidic acid acyltransferase, zeta)
Gene Aliases
AU041707, AW545732, GPAT4, Tsarg7
Chromosome Location
Chr.8: 23171265 - 23209171 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_018743.4 NP_061213.2 5 - 6 1575 85
GenBank mRNA AF015811.1 - 2 - 3 251 85
AF406611.1 - 4 - 5 1086 85
AK045235.1 - 5 - 6 1469 85
AK077561.1 - 5 - 6 1459 85
AK083589.1 - 5 - 6 1469 85
AK161270.1 - 5 - 6 1438 85
AK162033.1 - 2 - 3 380 85
AK167876.1 - 5 - 6 1484 85
AK169257.1 - 5 - 6 1469 85
AY489184.1 - 4 - 5 644 85
BC031767.1 - 4 - 5 929 85
BC057860.1 - 2 - 3 249 85

View Full Product Details