Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Casp8ap2
Entrez Gene ID
26885
Gene Name
caspase 8 associated protein 2
Gene Aliases
AA387232, D4Ertd659e, FLASH, mKIAA1315
Chromosome Location
Chr.4: 32615473 - 32653271 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001122978.1 NP_001116450.1 4 - 5 275 65
NM_011997.2 NP_036127.2 4 - 5 522 65
GenBank mRNA AF132726.1 - 4 - 5 482 65
AK142148.1 - 4 - 5 274 65
AK165109.1 - 4 - 5 272 65
BC138022.1 - 3 - 4 265 65
BC138041.1 - 3 - 4 265 65

View Full Product Details