The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_009138.3 | NP_033164.1 | 3 - 4 | 305 | 73 |
NR_033527.1 | 3 - 4 | 330 | 73 | ||
GenBank mRNA | AJ249480.1 | - | 3 - 4 | 278 | 73 |
AK131875.1 | - | 3 - 4 | 306 | 73 | |
AK132700.1 | - | 3 - 4 | 330 | 73 | |
AK134138.1 | - | 3 - 4 | 290 | 73 | |
BC117033.1 | - | 2 - 3 | 238 | 73 | |
BC117037.1 | - | 2 - 3 | 238 | 73 | |
DQ158256.1 | - | 3 - 4 | 291 | 73 | |
U86357.1 | - | 2 - 3 | 187 | 73 |