The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001007154.3 | NP_001007155.1 | 2 - 3 | 270 | 94 |
NM_001177789.1 | NP_001171260.1 | 2 - 3 | 488 | 94 | |
NM_001177790.1 | NP_001171261.1 | 2 - 3 | 488 | 94 | |
NM_001177791.1 | NP_001171262.1 | 2 - 3 | 322 | 94 | |
NM_028806.2 | NP_083082.1 | 2 - 3 | 727 | 94 | |
GenBank mRNA | AJ632220.1 | - | 2 - 3 | 285 | 94 |
AJ632221.1 | - | 2 - 3 | 323 | 94 | |
AJ632222.1 | - | 2 - 3 | 270 | 94 | |
AJ632223.1 | - | 1 - 2 | 160 | 94 | |
AK005136.1 | - | 2 - 3 | 292 | 94 | |
AK032242.1 | - | 2 - 3 | 494 | 94 | |
AK038449.1 | - | 2 - 3 | 238 | 94 | |
AK039213.1 | - | 2 - 3 | 277 | 94 | |
AK135144.1 | - | 2 - 3 | 728 | 94 | |
AK220554.1 | - | 2 - 3 | 488 | 94 | |
AY501387.1 | - | 2 - 3 | 434 | 94 | |
BC058621.1 | - | 2 - 3 | 643 | 94 | |
BC059869.1 | - | 2 - 3 | 506 | 94 | |
BY127674.1 | - | 2 - 3 | 271 | 94 |