The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001007154.3 | NP_001007155.1 | 3 - 4 | 351 | 75 |
NM_001177789.1 | NP_001171260.1 | 3 - 4 | 569 | 75 | |
NM_001177790.1 | NP_001171261.1 | 3 - 4 | 569 | 75 | |
NM_001177791.1 | NP_001171262.1 | 3 - 4 | 403 | 75 | |
NM_028806.2 | NP_083082.1 | 3 - 4 | 808 | 75 | |
GenBank mRNA | AJ632220.1 | - | 3 - 4 | 366 | 75 |
AJ632221.1 | - | 3 - 4 | 404 | 75 | |
AJ632222.1 | - | 3 - 4 | 351 | 75 | |
AJ632223.1 | - | 2 - 3 | 241 | 75 | |
AK005136.1 | - | 3 - 4 | 373 | 75 | |
AK032242.1 | - | 3 - 4 | 575 | 75 | |
AK038449.1 | - | 3 - 4 | 319 | 75 | |
AK039213.1 | - | 3 - 4 | 358 | 75 | |
AK135144.1 | - | 3 - 4 | 809 | 75 | |
AK220554.1 | - | 3 - 4 | 569 | 75 | |
AY501387.1 | - | 3 - 4 | 515 | 75 | |
BC058621.1 | - | 3 - 4 | 724 | 75 | |
BC059869.1 | - | 3 - 4 | 587 | 75 |