Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Pex11g
- Entrez Gene ID
-
69129
- Gene Name
- peroxisomal biogenesis factor 11 gamma
- Gene Aliases
-
1810022F11Rik, 1810049N02Rik, Pex11c, Pex11gamma
- Chromosome Location
-
Chr.8: 3457105 - 3467839 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK007848.1 |
- |
4 - 5 |
468 |
98 |
View Full Product Details