Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Wdr19
Entrez Gene ID
213081
Gene Name
WD repeat domain 19
Gene Aliases
C330027H04Rik, D330023L08Rik, DYF2, Ift144, PWDMP, mKIAA1638
Chromosome Location
Chr.5: 65199658 - 65260415 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_153391.2 NP_700440.2 10 - 11 1022 121
GenBank mRNA AK043836.1 - 10 - 11 979 121
AK052303.1 - 10 - 11 1002 121
AK129413.1 - 2 - 3 111 121
AK147970.1 - 10 - 11 1022 121
AK166172.1 - 10 - 11 988 121
AY029258.1 - 10 - 11 1092 121
BC060134.1 - 10 - 11 1016 121
BC082301.1 - 9 - 10 902 121

View Full Product Details