Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Stx18
Entrez Gene ID
71116
Gene Name
syntaxin 18
Gene Aliases
1810035L21Rik, 4933425D03Rik, AU041562
Chromosome Location
Chr.5: 38038622 - 38137770 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001289535.1 NP_001276464.1 2 - 3 339 96
NM_001289536.1 NP_001276465.1 2 - 3 339 96
NM_026959.3 NP_081235.2 2 - 3 339 96
GenBank mRNA AK007700.1 - 2 - 3 337 96
AK016910.1 - 2 - 3 281 96
AK169273.1 - 2 - 3 283 96
AK170165.1 - 2 - 3 335 96
BC021362.1 - 2 - 3 329 96

View Full Product Details