The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_016899.4 | NP_058595.2 | 4 - 5 | 714 | 63 |
GenBank mRNA | AB232620.1 | - | 4 - 5 | 510 | 63 |
AF119675.1 | - | 4 - 5 | 650 | 63 | |
AK003260.1 | - | 4 - 5 | 715 | 63 | |
BC006624.1 | - | 4 - 5 | 700 | 63 |