Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Nrbp1
Entrez Gene ID
192292
Gene Name
nuclear receptor binding protein 1
Gene Aliases
B230344L17Rik, Nrbp
Chromosome Location
Chr.5: 31228089 - 31251566 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_147201.2 NP_671734.1 6 - 7 669 65
GenBank mRNA AF302138.1 - 6 - 7 668 65
AK046142.1 - 6 - 7 663 65
AK077440.1 - 6 - 7 705 65
AK170501.1 - 6 - 7 685 65
AK218775.1 - 3 - 4 208 65
BC004756.1 - 6 - 7 639 65
BC018463.1 - 6 - 7 643 65

View Full Product Details