Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Lrtm1
Entrez Gene ID
319476
Gene Name
leucine-rich repeats and transmembrane domains 1
Gene Aliases
A930016D02Rik
Chromosome Location
Chr.14: 29018208 - 29033642 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_176920.4 NP_795894.1 1 - 2 395 136
GenBank mRNA AK044492.1 - 1 - 2 349 136
AK138618.1 - 1 - 2 213 136
BC099587.1 - 1 - 2 193 136
BC132594.1 - 1 - 2 99 136
BC145196.1 - 1 - 2 99 136
BC145990.1 - 1 - 2 99 136

View Full Product Details