Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Slitrk3
Entrez Gene ID
386750
Gene Name
SLIT and NTRK-like family, member 3
Gene Aliases
-
Chromosome Location
Chr.3: 73047247 - 73057881 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_198864.2 NP_942564.2 1 - 2 396 93
GenBank mRNA AK139378.1 - 1 - 2 520 93
BC066059.1 - 1 - 2 396 93

View Full Product Details