Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ythdc1
Entrez Gene ID
231386
Gene Name
YTH domain containing 1
Gene Aliases
A730098D12Rik, C80342, mKIAA1966
Chromosome Location
Chr.5: 86803980 - 86836659 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_177680.3 NP_808348.2 8 - 9 1311 142
GenBank mRNA AK173312.1 - 5 - 6 574 142
BC037593.1 - 2 - 3 204 142
BC044650.1 - 6 - 7 1129 142
CO424360.1 - 3 - 4 256 142

View Full Product Details