Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Vezt
Entrez Gene ID
215008
Gene Name
vezatin, adherens junctions transmembrane protein
Gene Aliases
A630071D13Rik, AI848282
Chromosome Location
Chr.10: 93961522 - 94035799 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK031848.1 - 7 - 8 1103 62
AK036017.1 - 7 - 8 1040 62
AK042540.1 - 7 - 8 1034 62
AY753561.1 - 7 - 8 996 62
AY753562.1 - 7 - 8 996 62
BC056428.1 - 7 - 8 1027 62

View Full Product Details