Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Plscr1
Entrez Gene ID
22038
Gene Name
phospholipid scramblase 1
Gene Aliases
MmTRA1a, MmTRA1b, Nor1, Tra1, Tra1a, Tra1b, Tras1, Tras2
Chromosome Location
Chr.9: 92250194 - 92272561 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_011636.2 NP_035766.2 3 - 4 271 111
GenBank mRNA AF159593.1 - 3 - 4 218 111
AK003628.1 - 3 - 4 199 111
AK149903.1 - 3 - 4 199 111
BC002017.1 - 3 - 4 182 111
CK788898.1 - 3 - 4 288 111

View Full Product Details