The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_175271.4 | NP_780480.2 | 1 - 2 | 371 | 67 |
GenBank mRNA | AK045289.1 | - | 1 - 2 | 341 | 67 |
AK051709.1 | - | 1 - 2 | 342 | 67 | |
BB619126.1 | - | 1 - 2 | 372 | 67 | |
BC052178.1 | - | 1 - 2 | 362 | 67 | |
BC069178.1 | - | 1 - 2 | 283 | 67 | |
BC138893.1 | - | 1 - 2 | 172 | 67 | |
BC138894.1 | - | 1 - 2 | 172 | 67 |