Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Sema4c
Entrez Gene ID
20353
Gene Name
sema domain, immunoglobulin domain (Ig), transmembrane domain (TM) and short cytoplasmic domain, (semaphorin) 4C
Gene Aliases
AI426163, M-Sema F, Semacl1, Semaf, Semai, sema I
Chromosome Location
Chr.1: 36548640 - 36558381 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK090073.1 - 14 - 15 1929 88
AK148298.1 - 14 - 15 1926 88
AK173247.1 - 15 - 16 2166 88
S79463.1 - 14 - 15 1780 88

View Full Product Details