Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Sema4c
- Entrez Gene ID
-
20353
- Gene Name
- sema domain, immunoglobulin domain (Ig), transmembrane domain (TM) and short cytoplasmic domain, (semaphorin) 4C
- Gene Aliases
-
AI426163, M-Sema F, Semacl1, Semaf, Semai, sema I
- Chromosome Location
-
Chr.1: 36548640 - 36558381 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK148298.1 |
- |
1 - 2 |
211 |
62 |
View Full Product Details