Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Kcnk7
Entrez Gene ID
16530
Gene Name
potassium channel, subfamily K, member 7
Gene Aliases
2310014G03Rik, Kcnk6, Kcnk8, Knot, Mlk3
Chromosome Location
Chr.19: 5704476 - 5707405 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_010609.2 NP_034739.2 2 - 3 717 104
GenBank mRNA AF012324.1 - 2 - 3 777 104
AF022820.1 - 2 - 3 714 104
AF110521.1 - 2 - 3 828 104
AK160180.1 - 2 - 3 724 104
BC106993.2 - 2 - 3 717 104

View Full Product Details