Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Acpp
- Entrez Gene ID
-
56318
- Gene Name
- acid phosphatase, prostate
- Gene Aliases
-
5'-NT, A030005E02Rik, FRAP, Lap, PAP, Ppal
- Chromosome Location
-
Chr.9: 104288240 - 104337722 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
BC139826.1 |
- |
1 - 2 |
169 |
59 |
View Full Product Details