The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_026417.4 | NP_080693.2 | 5 - 6 | 826 | 88 |
GenBank mRNA | AK017721.1 | - | 5 - 6 | 89 | |
AK017722.1 | - | 5 - 6 | 89 | ||
AK083301.1 | - | 5 - 6 | 806 | 88 | |
AK159227.1 | - | 5 - 6 | 787 | 88 | |
AK166347.1 | - | 5 - 6 | 814 | 88 | |
BC003936.1 | - | 6 - 7 | 1001 | 88 |