Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Wdr82
Entrez Gene ID
77305
Gene Name
WD repeat domain containing 82
Gene Aliases
9430077D24Rik, CDW5/WDR82
Chromosome Location
Chr.9: 106170929 - 106191706 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_029896.1 NP_084172.1 2 - 3 548 97
GenBank mRNA AK035109.1 - 2 - 3 550 97
AK037620.1 - 2 - 3 555 97
AK149692.1 - 2 - 3 549 97
BC019115.1 - 1 - 2 74 97
BC031502.1 - 2 - 3 238 97
BC157943.1 - - 471 97
BC157945.1 - - 472 97
BC158054.1 - - 472 97
EF011615.1 - 2 - 3 237 97

View Full Product Details