Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Pof1b
Entrez Gene ID
69693
Gene Name
premature ovarian failure 1B
Gene Aliases
2310066B14Rik
Chromosome Location
Chr.X: 112638427 - 112698711 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_181579.1 NP_853557.1 6 - 7 834 94
GenBank mRNA AF309775.1 - 5 - 6 727 94
AK036922.1 - 6 - 7 839 94
AK086511.1 - 6 - 7 835 94
AK137653.1 - 6 - 7 835 94

View Full Product Details