Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Lhcgr
Entrez Gene ID
16867
Gene Name
luteinizing hormone/choriogonadotropin receptor
Gene Aliases
Gpcr19-rs1, LH-R, Lhr
Chromosome Location
Chr.17: 88716488 - 88792059 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_013582.2 NP_038610.1 2 - 3 255 94
GenBank mRNA AK030325.1 - 2 - 3 270 94
M81310.1 - 2 - 3 255 94

View Full Product Details