Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Lhcgr
Entrez Gene ID
16867
Gene Name
luteinizing hormone/choriogonadotropin receptor
Gene Aliases
Gpcr19-rs1, LH-R, Lhr
Chromosome Location
Chr.17: 88716488 - 88792059 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_013582.2 NP_038610.1 5 - 6 472 92
GenBank mRNA AK030325.1 - 5 - 6 487 92
AK054331.1 - 4 - 5 309 92
AK138461.1 - 3 - 4 329 92
M81310.1 - 5 - 6 472 92

View Full Product Details