The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001039586.1 | NP_001034675.1 | 1 - 2 | 98 | 78 |
GenBank mRNA | AF455785.1 | - | 1 - 2 | 67 | 78 |
AK050182.1 | - | 1 - 2 | 88 | 78 | |
BC036136.1 | - | - | 55 | 78 | |
BY111608.1 | - | 1 - 2 | 99 | 78 |