Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Socs5
- Entrez Gene ID
-
56468
- Gene Name
- suppressor of cytokine signaling 5
- Gene Aliases
-
1810018L08Rik, Cish5, mKIAA0671
- Chromosome Location
-
Chr.17: 87107407 - 87137647 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK135099.1 |
- |
1 - 2 |
356 |
72 |
View Full Product Details