Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Rcor2
Entrez Gene ID
104383
Gene Name
REST corepressor 2
Gene Aliases
1A13, AW122124, CoREST, Rcor, Rcor1
Chromosome Location
Chr.19: 7267330 - 7275225 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_054048.3 NP_473389.1 7 - 8 767 63
GenBank mRNA AK052295.1 - 9 - 10 1021 63
BC055719.1 - 8 - 9 1188 63
X83587.2 - 7 - 8 769 63

View Full Product Details