Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Nrxn2
Entrez Gene ID
18190
Gene Name
neurexin II
Gene Aliases
6430591O13Rik, mKIAA0921
Chromosome Location
Chr.19: 6418738 - 6533217 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001205234.1 NP_001192163.1 8 - 9 1815 75
NM_001205235.1 NP_001192164.1 7 - 8 1791 75
NM_020253.3 NP_064649.2 8 - 9 1815 75
GenBank mRNA AK129239.1 - 8 - 9 1788 75
AK147295.1 - 8 - 9 1750 75
AK147665.1 - 8 - 9 1750 75
AK170622.1 - 2 - 3 871 75
BC145496.1 - 6 - 7 1438 75
BC150784.1 - 6 - 7 1438 75

View Full Product Details