The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_207268.3 | NP_997151.2 | 1 - 1 | 3012 | 131 |
GenBank mRNA | BB667164.1 | - | 1 - 1 | 518 | 131 |