Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cdc25a
Entrez Gene ID
12530
Gene Name
cell division cycle 25A
Gene Aliases
D9Ertd393e
Chromosome Location
Chr.9: 109875337 - 109893895 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_007658.3 NP_031684.3 10 - 11 1406 73
GenBank mRNA AK077609.1 - 10 - 11 1024 73
AK077991.1 - 10 - 11 1409 73
AK142334.1 - 9 - 10 1681 73
BC046296.1 - 10 - 11 1025 73

View Full Product Details