Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cdc25a
Entrez Gene ID
12530
Gene Name
cell division cycle 25A
Gene Aliases
D9Ertd393e
Chromosome Location
Chr.9: 109875337 - 109893895 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_007658.3 NP_031684.3 2 - 3 654 62
GenBank mRNA AK077609.1 - 2 - 3 271 62
AK077991.1 - 2 - 3 657 62
AK142334.1 - 1 - 2 929 62
BC046296.1 - 2 - 3 273 62
U27323.1 - 2 - 3 240 62

View Full Product Details