The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_030563.2 | NP_085040.2 | 6 - 7 | 2568 | 79 |
GenBank mRNA | AK147522.1 | - | 6 - 7 | 2569 | 79 |
AK172981.1 | - | 6 - 7 | 2550 | 79 | |
BC004022.1 | - | 3 - 4 | 1115 | 79 | |
BC089324.1 | - | 5 - 6 | 1733 | 79 | |
BC111031.1 | - | 2 - 3 | 2537 | 79 |