The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001037754.3 | NP_001032843.1 | 1 - 2 | 160 | 76 |
NM_001037755.3 | NP_001032844.2 | 1 - 2 | 160 | 76 | |
NM_130862.4 | NP_570932.2 | 1 - 2 | 160 | 76 | |
GenBank mRNA | AF390178.1 | - | 1 - 2 | 118 | 76 |
AF390179.1 | - | 1 - 2 | 118 | 76 | |
AK049469.1 | - | 1 - 2 | 161 | 76 | |
AK145924.1 | - | 1 - 2 | 161 | 76 | |
AK160401.1 | - | 1 - 2 | 161 | 76 | |
AK170190.1 | - | 1 - 2 | 163 | 76 | |
BC006620.1 | - | 1 - 2 | 151 | 76 | |
BC016411.1 | - | 1 - 2 | 151 | 76 |