Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Aatk
Entrez Gene ID
11302
Gene Name
apoptosis-associated tyrosine kinase
Gene Aliases
AATYK, aatyk1, mKIAA0641
Chromosome Location
Chr.11: 120007311 - 120047182 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001198785.1 NP_001185714.1 5 - 6 763 81
NM_001198787.1 NP_001185716.1 5 - 6 535 81
NM_007377.4 NP_031403.2 5 - 6 1046 81
GenBank mRNA AB093253.2 - 5 - 6 1046 81
AB288871.1 - 5 - 6 665 81
AF011908.1 - 5 - 6 502 81
AK036705.1 - 5 - 6 537 81
AK147239.1 - 2 - 3 556 81
AK155100.1 - 5 - 6 537 81
AY236858.1 - 5 - 6 541 81
AY236859.1 - 5 - 6 568 81
BC042837.1 - 5 - 6 1068 81
BC052323.1 - 5 - 6 1068 81
BC080846.1 - 5 - 6 617 81

View Full Product Details