Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Prss42
Entrez Gene ID
235628
Gene Name
protease, serine 42
Gene Aliases
Tessp2
Chromosome Location
Chr.9: 110797261 - 110803744 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_153099.1 NP_694739.1 1 - 2 221 73
GenBank mRNA AB052292.1 - 1 - 2 221 73
AK133402.1 - 1 - 2 86 73
BC060984.1 - 1 - 2 240 73

View Full Product Details