The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_153099.1 | NP_694739.1 | 3 - 4 | 647 | 148 |
GenBank mRNA | AB052292.1 | - | 3 - 4 | 647 | 148 |
AK133402.1 | - | 3 - 4 | 512 | 148 | |
BC060984.1 | - | 3 - 4 | 666 | 148 |