The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_009708.1 | NP_033838.1 | 4 - 5 | 433 | 66 |
GenBank mRNA | AF016482.1 | - | 4 - 5 | 416 | 66 |
AK075970.1 | - | - | 741 | 66 | |
AK170529.1 | - | - | 1864 | 66 | |
BC107022.2 | - | 4 - 5 | 432 | 66 | |
BC127034.1 | - | 4 - 5 | 427 | 66 | |
BC128475.1 | - | 4 - 5 | 432 | 66 | |
BC139330.1 | - | 4 - 5 | 506 | 66 | |
BC139331.1 | - | 4 - 5 | 506 | 66 |