Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Nol11
Entrez Gene ID
68979
Gene Name
nucleolar protein 11
Gene Aliases
1500002M01Rik, AU015220, AU019874
Chromosome Location
Chr.11: 107166660 - 107189381 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001161329.1 NP_001154801.1 13 - 14 1645 102
GenBank mRNA AK038696.1 - 13 - 14 1536 102
AK076704.1 - 13 - 14 1646 102

View Full Product Details