Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Nol11
Entrez Gene ID
68979
Gene Name
nucleolar protein 11
Gene Aliases
1500002M01Rik, AU015220, AU019874
Chromosome Location
Chr.11: 107166660 - 107189381 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001161329.1 NP_001154801.1 6 - 7 793 106
NM_133702.2 NP_598463.2 6 - 7 793 106
GenBank mRNA AK005123.1 - 6 - 7 683 106
AK038696.1 - 6 - 7 684 106
AK054118.1 - 2 - 3 1399 106
AK076704.1 - 6 - 7 794 106
AK078524.1 - 6 - 7 684 106
AK157743.1 - 6 - 7 733 106

View Full Product Details