Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Kcnb1
- Entrez Gene ID
-
16500
- Gene Name
- potassium voltage gated channel, Shab-related subfamily, member 1
- Gene Aliases
-
Kcr1-1, Kv2.1, Shab
- Chromosome Location
-
Chr.2: 167095969 - 167188818 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
BC054394.1 |
- |
2 - 3 |
528 |
82 |
View Full Product Details