Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Spata2
Entrez Gene ID
263876
Gene Name
spermatogenesis associated 2
Gene Aliases
AI504642, mKIAA0757
Chromosome Location
Chr.2: 167481136 - 167492874 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK144826.1 - 3 - 4 100

View Full Product Details